Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZAP70 Rabbit anti-Human, Mouse, Unconjugated, Clone: H11, Abnova™
Rabbit Monoclonal Antibody.
Specifications
Antigen | ZAP70 |
---|---|
Clone | H11 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-336-002
|
Abnova Corporation
MAB19168 |
20 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
89-336-001
|
Abnova Corporation
MAB19167 |
200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
ZAP70 Monoclonal antibody specifically detects ZAP70 in Human, Mouse samples. It is validated for WB-Ce, Flow Cyt.Specifications
ZAP70 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
FLJ17670, FLJ17679, SRK, STD, TZK, ZAP-70 | |
A synthetic phosphopeptide corresponding to residues surrounding Y493 of human ZAP70 | |
Primary | |
zeta-chain (TCR) associated protein kinase 70kDa |
H11 | |
Monoclonal | |
Liquid | |
Human, Mouse | |
7535, 6850 | |
IgG1 κ | |
Store at -20°C | |
ZAP70 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title