Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAPA sulfate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

371962-01-5
C4H8N2O6S2
244.236
UWVNHPNVOMFDHW-ODZAUARKSA-N
(Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
90479730

Agonist at 'low affinity' GABAA receptor. More potent than GABA/muscimol

More potent than either GABA or muscimol as an agonist at low affinity GABAA receptors and is thus a useful ligand to investigate GABA receptors linked to benzodiazepine receptors. Also a GABAA-ρ receptor antagonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C(=CSC(=N)N)C(=O)O.OS(=O)(=O)O
IUPAC Name (Z)-3-carbamimidoylsulfanylprop-2-enoic acid;sulfuric acid
Formula Weight 244.24
Chemical Name or Material ZAPA sulfate
Grade >98%

Recommended Storage : Store at Room Temperature