Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAPA sulfate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

371962-01-5
C4H8N2O6S2
244.236
UWVNHPNVOMFDHW-ODZAUARKSA-N
(Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
90479730

Agonist at 'low affinity' GABAA receptor. More potent than GABA/muscimol

More potent than either GABA or muscimol as an agonist at low affinity GABAA receptors and is thus a useful ligand to investigate GABA receptors linked to benzodiazepine receptors. Also a GABAA-ρ receptor antagonist.
CAS 371962-01-5
Molecular Formula C4H8N2O6S2
Quantity 50mg
Synonym (Z)-3-[(Aminoiminomethyl)thio]prop-2-enoic acid sulfate
InChI Key UWVNHPNVOMFDHW-ODZAUARKSA-N
SMILES C(=CSC(=N)N)C(=O)O.OS(=O)(=O)O
IUPAC Name (Z)-3-carbamimidoylsulfanylprop-2-enoic acid;sulfuric acid
Molecular Weight (g/mol) 244.236
PubChem CID 90479730
Formula Weight 244.24
Chemical Name or Material ZAPA sulfate
Grade >98%
Show More Show Less

Recommended Storage : Store at Room Temperature