Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548591

Catalog No. PA548591

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZBTB11
Polyclonal
PBS with 0.09% sodium azide
O95625
Zinc finger and BTB domain-containing protein 11, ZBTB11
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27107
Western Blot
Unconjugated
ZBTB11
Liquid
ZBTB11
KLH conjugated synthetic peptide between 736-765 amino acids from the C-terminal region of human ZBTB11.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit