Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB20 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ZBTB20
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095891
View Documents
Novus Biologicals
NBP304370100UL
100 μg Each for $320.71
Add to cart
 
NB095892
View Documents
Novus Biologicals
NBP30437020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ZBTB20 Polyclonal antibody specifically detects ZBTB20 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

ZBTB20
Unconjugated
Rabbit
Human
BTB/POZ zinc finger protein DPZF, dendritic cell-derived BTB/POZ zinc finger, Dendritic-derived BTB/POZ zinc finger protein, DPZFZinc finger protein 288ZNF288DKFZp566F123, FLJ26458, HOF, ODA-8S, zinc finger and BTB domain containing 20, zinc finger and BTB domain-containing protein 20
Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human ZBTB20 (NP_056457.3).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
26137
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit