Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB26 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen ZBTB26
Immunogen Synthetic peptide directed towards the C terminal of human ZBTB26. Peptide sequence VFAHKPVLRKHLKQLHGKNSFDNANERNVQDLTVDFDSFACTTVTDSKGC.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1803520 View Documents Novus Biologicals
NBP18035120UL
20ul Each for $152.22
Add to cart
 
NBP180351 View Documents Novus Biologicals
NBP180351
100 ul Each for $378.35
Add to cart
 
Description

Description

ZBTB26 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZBTB26
Rabbit
Polyclonal
Unconjugated
57684
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the C terminal of human ZBTB26. Peptide sequence VFAHKPVLRKHLKQLHGKNSFDNANERNVQDLTVDFDSFACTTVTDSKGC.
Primary
IgG
KIAA1572, zinc finger and BTB domain containing 26, zinc finger and BTB domain-containing protein 26, Zinc finger protein 481bioref, zinc finger protein 483, Zinc finger protein Bioref, ZNF481
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit