Learn More
Abnova™ ZBTB3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00079842T01
Catalog No. 89-967-694
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZBTB3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 63.25kDa
Transient overexpression cell lysate was tested with anti-ZBTB3 antibody (Mfr. No. H00079842-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_024784.2
pCMV-ZBTB3 full-length
100 μL
Human