Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176516MFV610
Description
ZBTB3 Polyclonal antibody specifically detects ZBTB3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ZBTB3 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ23392, zinc finger and BTB domain containing 3, zinc finger and BTB domain-containing protein 3 | |
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZBTB3. The immunogen is located within amino acids 510 - 560 of ZBTB3. Amino Acid Squence: RKAHNEDLAKRSKPDPE | |
0.1 mL | |
Signal Transduction | |
79842 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction