Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB37 Mouse anti-Human, DyLight 488, Clone: OTI1D7, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274907G
Description
ZBTB37 Monoclonal antibody specifically detects ZBTB37 in Human samples. It is validated for Western Blot.Specifications
ZBTB37 | |
OTI1D7 | |
Western Blot | |
Affinity Purified, Conjugated | |
ZBTB37 | |
Recombinant protein fragment corresponding to amino acids 131-361 of human ZBTB37 (NP_115911) produced in E.coli. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
D430004I08Rik, MGC2629, zinc finger and BTB domain containing 37, zinc finger and BTB domain-containing protein 37, ZNF908 | |
Mouse | |
IgG2b | |
0.1 mL | |
Primary | |
84614.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title