Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB37 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA803616
Description
ZBTB37 may be involved in transcriptional regulation.Specifications
ZBTB37 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5TC79 | |
ZBTB37 | |
Human recombit protein fragment corresponding to amino acids 131-361 of human ZBTB37 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI1D7 | |
Unconjugated | |
ZBTB37 | |
D430004I08Rik, ZNF908 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84614 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction