Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB37 Mouse anti-Human, DyLight™ 350, Clone: OTI1D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274907UV

Catalog No. NBP274907UV


Add to cart

Description

Description

ZBTB37 Monoclonal antibody specifically detects ZBTB37 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZBTB37
OTI1D7
Western Blot
Affinity Purified, Conjugated
ZBTB37
Recombinant protein fragment corresponding to amino acids 131-361 of human ZBTB37 (NP_115911) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
D430004I08Rik, MGC2629, zinc finger and BTB domain containing 37, zinc finger and BTB domain-containing protein 37, ZNF908
Mouse
IgG2b
0.1 mL
Primary
84614.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit