Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB42 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548243

Catalog No. PA548243

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcriptional repressor. Specifically binds DNA and probably acts by recruiting chromatin remodeling multiprotein complexes.
Specifications

Specifications

ZBTB42
Polyclonal
PBS with 0.09% sodium azide
B2RXF5, Q811H0
Zinc finger and BTB domain-containing protein 42, ZBTB42
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100128927, 382639
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZBTB42
Liquid
ZBTB42
KLH conjugated synthetic peptide between 37-65 amino acids from the N-terminal region of human ZBTB42.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit