Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB44 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548835

Catalog No. PA548835

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

May be involved in transcriptional regulation.
Specifications

Specifications

ZBTB44
Polyclonal
PBS with 0.09% sodium azide
Q8NCP5
Zinc finger and BTB domain-containing protein 44, BTB/POZ domain-containing protein 15, Zinc finger protein 851, ZBTB44, BTBD15, ZNF851
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29068
Western Blot
Unconjugated
ZBTB44
Liquid
ZBTB44
KLH conjugated synthetic peptide between 425-453 amino acids from the C-terminal region of human ZBTB44.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit