Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA573021
Description
ZBTB46 Polyclonal Antibody for ELISA
ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
Specifications
ZBTB46 | |
Polyclonal | |
Unconjugated | |
ZBTB46 | |
2610019F01Rik; 4933406L05Rik; BTB (POZ) domain containing 4; BTB/POZ domain-containing protein 4; Btbd4; BTB-ZF protein expressed in effector lymphocytes; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; zinc finger and BTB domain containing 46; zinc finger and BTB domain-containing protein 46; Zinc finger protein 340; ZNF340 | |
Rabbit | |
Protein G | |
RUO | |
140685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q86UZ6 | |
ZBTB46 | |
Amino acids 62-87 of human RINZF protein were used as the immunogen. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction