Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZBTB7C Mouse anti-Human, Clone: PCRP-ZBTB7C-4E12, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ZBTB7C
Clone PCRP-ZBTB7C-4E12
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121778
View Documents
Novus Biologicals
NBP30715220UG
20 μg
Each for $138.37
Add to cart
 
NB121777
View Documents
Novus Biologicals
NBP30715210UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ZBTB7C Monoclonal antibody specifically detects ZBTB7C in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

ZBTB7C
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
201501
IgG2b
Primary
Store at 4°C.
PCRP-ZBTB7C-4E12
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Human
affected by papillomavirus DNA integration in ME180 cells protein 1, APM1, APM-1, B230208J24Rik, FLJ37907, ZBTB36, zinc finger and BTB domain containing 36, zinc finger and BTB domain containing 7C, zinc finger and BTB domain-containing protein 36, zinc finger and BTB domain-containing protein 7C, zinc finger protein 857C, ZNF857C
Recombinant full-length human ZBTB7C protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit