Learn More
Invitrogen™ ZBTB8A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116783
Description
Positive Samples: U-87MG, PC-3, HeLa, Mouse lung, Mouse brain, Rat heart Immunogen sequence: HEPRKESIKK TKHLRLSQPS EVTHYKSSKR EVRTSDSSSH VSQSEEQAQI DAEMDSTPVG YQYGQGSDVT SKSFPDDLPR MRFKCPYCTH VVKRKADLKR HLRCHTGERP YPCQACGKRF SRLDHLSSHF RTIHQACKLI CRKCKRHVTD LTGQVVQEGT RRYRLCNECL AEFGIDSLPI DLEAEQHLMS PSDGDKDSRW H.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB8, also known as BOZ-F1, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB8, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB8 may act as a transcriptional repressor or be involved in tumorigenesis.
Specifications
ZBTB8A | |
Polyclonal | |
Unconjugated | |
ZBTB8A | |
2410081M15Rik; AI480941; BOZF1; BOZ-F1; BTB/POZ and zinc-finger domain-containing factor; BTB/POZ and zinc-finger domains factor on chromosome 1; ZBTB8; Zbtb8a; zinc finger and BTB domain containing 8; zinc finger and BTB domain containing 8a; zinc finger and BTB domain-containing protein 8A; ZNF916A | |
Rabbit | |
Affinity chromatography | |
RUO | |
313049, 653121, 73680 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.82 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
B1WBU4, Q96BR9, Q9CWH1 | |
ZBTB8A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human ZBTB8A (NP_001035531). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.