Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537081
Description
This antibody detects endogenous protein at a molecular weight of 89 kDa. Purity is >95% by SDS-PAGE.
Involved in nuclear mRNA export; probably mediated by association with the TREX complex. [UniProt].
Specifications
ZC3H11A | |
Polyclonal | |
Unconjugated | |
ZC3H11A | |
1110003F06Rik; 5730454B08Rik; G630041M05Rik; Kiaa0663; mKIAA0663; RGD1308290; ZC3H11A; ZC3HDC11A; Zc3hh11a; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH type containing 11A; zinc finger CCCH-type containing 11A; zinc finger CCCH-type domain containing 11A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360845, 70579, 9877 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O75152, Q6NZF1 | |
ZC3H11A | |
Synthetic peptide corresponding to amino acids 339-384 of human ZC3H11A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction