Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H12D Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281878PCP
Description
ZC3H12D Polyclonal antibody specifically detects ZC3H12D in Human samples. It is validated for Western Blot, ELISASpecifications
| ZC3H12D | |
| Polyclonal | |
| Western Blot, ELISA | |
| C6orf95, FLJ46041, MCP-induced protein 4, p34, TFL, transformed follicular lymphoma, tumor suppressor TFL, zinc finger CCCH-type containing 12D | |
| ZC3H12D antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZC3H12D. The immunogen is located within amino acids 280 - 330 of ZC3H12D. Amino Acid Squence: TGARPGAGAEEQRPPRAPG | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 340152 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction