Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556638
Description
Immunogen sequence: AWKKRKRQEK IDKLEQDMER RKADFKAGKA LVISGREVFE FRPELVNDDD EEADDTRYTQ GTGGDEVDDS VSVND Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
Protects DRG1 from proteolytic degradation (PubMed:19819225). Stimulates DRG1 GTPase activity likely by increasing the affinity for the potassium ions (PubMed:23711155). [UniProt].
Specifications
ZC3H15 | |
Polyclonal | |
Unconjugated | |
Zc3h15 | |
1700006A17Rik; 1810012H02Rik; 2610312B22Rik; brain zinc finger protein; DFRP1; DRG family-regulatory protein 1; epo-immediate response gene protein FM22; FM22; HSPC303; HT010; Ierepo4; LEREPO4; likely ortholog of mouse immediate early response erythropoietin 4; MSTP012; p48ZnF; PP730; Zc3h15; Zinc finger CCCH domain-containing protein 15; zinc finger CCCH-type containing 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362154, 55854, 69082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3TIV5, Q6U6G5, Q8WU90 | |
Zc3h15 | |
Recombinant protein corresponding to Human ZC3H15. Recombinant protein control fragment (Product #RP-95965). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction