Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H7A Mouse anti-Human, DyLight 680, Clone: PCRP-ZC3H7A-1D6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314047FR
This item is not returnable.
View return policy
Description
ZC3H7A Monoclonal antibody specifically detects ZC3H7A in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZC3H7A | |
Monoclonal | |
DyLight 680 | |
FLJ10027, FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH domain-containing protein 7A, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type containing 7A, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807 | |
Recombinant full-length human ZC3H7A protein (Uniprot: Q8IWR0) | |
0.1 mL | |
Cell Biology | |
29066 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZC3H7A-1D6 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title