Learn More
ZC3H8 Mouse anti-Human, Clone: OTI4H5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808289
Catalog No. 50-166-8756
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZC3H8 acts as a transcriptional repressor of the GATA3 promoter. Sequence-specific DNA-binding factor that binds to the 5 ft.-AGGTCTC-3 ft. sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene. Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III. Induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.
Specifications
Specifications
ZC3H8
Monoclonal
Unconjugated
ZC3H8
Lyophilized
ZC3H8
Full length human recombit protein of human ZC3H8 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI4H5
PBS with 8% trehalose and no preservative; pH 7.3
Q8N5P1
Fliz1, ZC3HDC8
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
84524