Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548207
Description
Acts as a transcriptional repressor of the GATA3 promoter. Sequence-specific DNA-binding factor that binds to the 5′-AGGTCTC-3′ sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene. Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:23932780). Induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.Specifications
ZC3H8 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8N5P1, Q9JJ48 | |
ZC3H8 | |
KLH conjugated synthetic peptide between 52-81 amino acids from the N-terminal region of human ZC3H8. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
ZC3H8 | |
Zinc finger CCCH domain-containing protein 8, ZC3H8, ZC3HDC8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57432, 84524 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction