Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3HAV1L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549243

Catalog No. PA549243

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ZC3HAV1L
Polyclonal
PBS with 0.09% sodium azide
Q96H79
Zinc finger CCCH-type antiviral protein 1-like, ZC3HAV1L, C7orf39
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
92092
Western Blot
Unconjugated
ZC3HAV1L
Liquid
ZC3HAV1L
KLH conjugated synthetic peptide between 226-254 amino acids from the C-terminal region of human ZC3HAV1L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit