Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548897

Catalog No. PA548897

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcriptional coactivator in the bone morphogenetic protein (BMP)-signaling pathway. It positively modulates BMP signaling by interacting with SMAD1 and associating with CBP in the transcription complex. It contributes to the BMP-induced enhancement of cholinergic-neuron-specific gene expression.
Specifications

Specifications

ZCCHC12
Polyclonal
PBS with 0.09% sodium azide
Q6PEW1
Zinc finger CCHC domain-containing protein 12, Smad-interacting zinc finger protein 1, ZCCHC12, SIZN1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170261
Western Blot
Unconjugated
ZCCHC12
Liquid
ZCCHC12
KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human ZCCHC12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit