Learn More
Aviva Systems Biology Corp ZCCHC13 Antibody - middle region (ARP55945_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55945_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZCCHC13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZCCHC13. Alternate names: Cnbp2, ZNF9L, CNBP2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61671