Learn More
ZCCHC13 Mouse anti-Human, Clone: 4G11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524539
Catalog No. PIMA524539
Add to cart
Description
Description
Peptide Sequence: MSSKDFFACG HSGHWARGCP RGGAGGRRGG GHGRGSQCGS TTLSYTCYCC GESGRNAKNC VLLGNICYNC GRSGHIAKDC KDPKRERRQH CYTCGRLGHL ARDCDRQKEQ KCYSCGKLGH IQKDCAQVKC YRCGEIGHVA INCSKARPGQ LLPLRQIPTS SQGMSQ
This gene appears to represent an intronless retrocopy of a related multi-exon gene located on chromosome 3. However, the CDS of this intronless gene remains relatively intact, it is conserved in other mammalian species, it is known to be transcribed, and it is therefore thought to encode a functional protein. The encoded protein contains six CCHC-type zinc fingers, and is thus thought to function as a transcription factor. [provided by RefSeq, May 2010].
Specifications
Specifications
ZCCHC13
Monoclonal
Unconjugated
ZCCHC13
Liquid
ZCCHC13
ZCCHC13 (NP_976048.1, 1 a.a. ∽ 166 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
4G11
PBS with no preservative; pH 7.4
Q8WW36
ZCCHC13
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
389874