Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC24 Mouse anti-Human, DyLight™ 488, Clone: OTI2E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274909G

Catalog No. NBP274909G


Add to cart

Description

Description

ZCCHC24 Monoclonal antibody specifically detects ZCCHC24 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZCCHC24
OTI2E1
Western Blot
Affinity Purified, Conjugated
ZCCHC24
Full length recombinant protein of human ZCCHC24 (NP_699198) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
C10orf56, chromosome 10 open reading frame 56, FLJ90798, zinc finger CCHC domain-containing protein 24, zinc finger, CCHC domain containing 24
Mouse
IgG1
0.1 mL
Primary
219654.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit