Tocris™ ZD 2079

beta3-adrenoceptor agonist

Manufacturer: Tocris™ 2154/10

 View more versions of this product

Catalog No. 21-541-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors ZD 2079
    Purity >99
    Molecular Weight 351.83
    Quantity 10mg
    Product Type ZD 2079

    β3-adrenoceptor agonist. Relaxes rat mesenteric artery and isolated aorta in vitro. Inhibits ob gene expression and circulating leptin levels in lean mice in vivo.

    • Chemical Name: 4-[2-[[(2R)-2-Hydroxy-2-phenylethyl]amino]ethoxy]-benzeneacetic acid hydrochloride
    • Formula: C18H21NO4.HCl
    • CAS Number: 178600-17-4
    • Solubility: Soluble to 25mM in water
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit