Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZD 7114 hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

129689-28-7
C22H31ClN2O6
454.948
KCAMZVQSGDBGGF-FERBBOLQSA-N
(S)-4-[2-Hydroxy-3-phenoxypropylaminoethoxy]-N-(2-methoxyethyl)phenoxyacetamide hydrochloride
9890216

β3 agonist

β3-adrenoceptor agonist. Active metabolite ICI 215,001 hydrochloride (Cat. No. 0929) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES COCCNC(=O)COC1=CC=C(C=C1)OCCNCC(COC2=CC=CC=C2)O.Cl
IUPAC Name 2-[4-[2-[[(2S)-2-hydroxy-3-phenoxypropyl]amino]ethoxy]phenoxy]-N-(2-methoxyethyl)acetamide;hydrochloride
Formula Weight 454.95
Chemical Name or Material ZD 7114 hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature