Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZD 7114 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
129689-28-7
C22H31ClN2O6
454.948
KCAMZVQSGDBGGF-FERBBOLQSA-N
zd 7114 hydrochloride,zd7114 hydrochloride,d-7114 hydrochloride,zd-7114 hydrochloride,zd 7114 hydrochloride hplc,s-4-2-hydroxy-3-phenoxypropylaminoethoxy-n-2methoxyethyl phenoxyacetamide hydrochloride,s-2-4-2-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl acetamide hydrochloride,s-4-2-2-hydroxy-3-phenoxypropylamino ethoxy-n-2-methoxyethyl phenoxyacetamide hydrochloride,2-4-2-2s-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl acetamide hydrochloride,acetamide, 2-4-2-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl-, monohydrochloride, s
9890216
2-[4-[2-[[(2S)-2-hydroxy-3-phenoxypropyl]amino]ethoxy]phenoxy]-N-(2-methoxyethyl)acetamide;hydrochloride
COCCNC(=O)COC1=CC=C(C=C1)OCCNCC(COC2=CC=CC=C2)O.Cl
129689-28-7
C22H31ClN2O6
454.948
KCAMZVQSGDBGGF-FERBBOLQSA-N
zd 7114 hydrochloride,zd7114 hydrochloride,d-7114 hydrochloride,zd-7114 hydrochloride,zd 7114 hydrochloride hplc,s-4-2-hydroxy-3-phenoxypropylaminoethoxy-n-2methoxyethyl phenoxyacetamide hydrochloride,s-2-4-2-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl acetamide hydrochloride,s-4-2-2-hydroxy-3-phenoxypropylamino ethoxy-n-2-methoxyethyl phenoxyacetamide hydrochloride,2-4-2-2s-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl acetamide hydrochloride,acetamide, 2-4-2-2-hydroxy-3-phenoxypropyl amino ethoxy phenoxy-n-2-methoxyethyl-, monohydrochloride, s
9890216
2-[4-[2-[[(2S)-2-hydroxy-3-phenoxypropyl]amino]ethoxy]phenoxy]-N-(2-methoxyethyl)acetamide;hydrochloride
COCCNC(=O)COC1=CC=C(C=C1)OCCNCC(COC2=CC=CC=C2)O.Cl

β3 agonist

β3-adrenoceptor agonist. Active metabolite ICI 215,001 hydrochloride (Cat. No. 0929) also available.
Quantity 50mg
Formula Weight 454.95
Percent Purity >98%
Chemical Name or Material ZD 7114 hydrochloride

Recommended Storage : Store at Room Temperature