Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZD 7155 hydrochloride

Selective non-peptide AT1 antagonist

$187.00 - $787.00

Specifications

Assay Percent Range >98%
Formula Weight Observed MW: 474.99
Chemical Name or Material 5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride
Molecular Formula C26H26N6O.HCl
CAS 146709-78-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-111-0R
View Documents
Tocris Bioscience™
1211/10
10mg
Each for $187.00
Add to cart
 
12-115-0R
View Documents
Tocris Bioscience™
1211/50
50mg
Each for $787.00
Add to cart
 
Description

Description

A potent and selective competitive antagonist for the angiotensin II type 1 (AT1) receptor. Displaces [125I]-angiotensin II binding with an IC50 value of 3.8 nM in guinea pig adrenal gland membranes. Orally active, and is more potent and longer-acting than the prototype AT1 antagonist, losartan.
Specifications

Specifications

>98%
5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride
146709-78-6
Observed MW: 474.99
C26H26N6O.HCl
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at +4°C

SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit