Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548736

Catalog No. PA548736

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Palmitoyltransferase with broad specificity. Palmitoylates GABA receptors on their gamma subunit (GABRG1, GABRG2 and GABRG3), which regulates synaptic clustering and/or cell surface stability. Palmitoylates glutamate receptors GRIA1 and GRIA2, which leads to their retention in Golgi.
Specifications

Specifications

ZDHC3
Polyclonal
PBS with 0.09% sodium azide
Q9NYG2
Palmitoyltransferase ZDHHC3, Protein DHHC1, Zinc finger DHHC domain-containing protein 3, DHHC-3, Zinc finger protein 373, ZDHHC3, ZNF373
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51304
Western Blot
Unconjugated
ZDHHC3
Liquid
ZDHHC3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ZDHC3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit