Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ZDHHC13 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15902620
![]() |
Novus Biologicals
NBP15902620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159026
![]() |
Novus Biologicals
NBP159026 |
100 μL |
Each for $487.50
|
|
|||||
Description
ZDHHC13 Polyclonal specifically detects ZDHHC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZDHHC13 | |
Polyclonal | |
Purified | |
RUO | |
Q8IUH4-3 | |
54503 | |
Synthetic peptides corresponding to ZDHHC13(zinc finger, DHHC-type containing 13) The peptide sequence was selected from the N terminal of ZDHHC13. Peptide sequence MVILLLQHGADPTLIDGEGFSSIHLAVLFQHMPIIAYLISKGQSVNMTDV. | |
Primary |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
DHHC-13, EC 2.3.1, EC 2.3.1.-, FLJ10852, FLJ10941, HIP14LHIP14-related protein, HIP3RP, huntingtin interacting protein HIP3RP, Huntingtin-interacting protein 14-related protein, Huntingtin-interacting protein HIP3RP, MGC64994, palmitoyltransferase ZDHHC13, probable palmitoyltransferase ZDHHC13, Putative MAPK-activating protein PM03, Putative NF-kappa-B-activating protein 209, Zinc finger DHHC domain-containing protein 13, zinc finger, DHHC domain containing 13, zinc finger, DHHC-type containing 13 | |
ZDHHC13 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title