Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen ZDHHC16
Immunogen Synthetic peptides corresponding to ZDHHC16(zinc finger, DHHC-type containing 16) The peptide sequence was selected from the C terminal of ZDHHC16. Peptide sequence VLISRGETSIERHINKKERRRLQAKGRVFRNPYNYGCLDNWKVFLGVDTG.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15973720 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15973720UL
20ul Each for $149.00
Add to cart
 
NBP159737 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159737
100 ul Each for $329.00
Add to cart
 
Description

Description

ZDHHC16 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZDHHC16
Rabbit
Polyclonal
Unconjugated
84287
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to ZDHHC16(zinc finger, DHHC-type containing 16) The peptide sequence was selected from the C terminal of ZDHHC16. Peptide sequence VLISRGETSIERHINKKERRRLQAKGRVFRNPYNYGCLDNWKVFLGVDTG.
Primary
IgG
DHHC domain containing 16, EC 2.3.1, EC 2.3.1.-, zinc finger, DHHC-type containing 16
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit