Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZDHHC8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548795
Description
Predicted to react with bovine, mouse, rabbit and rat based on sequence homology.
This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.Specifications
ZDHHC8 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9ULC8, Q5Y5T5 | |
ZDHHC8 | |
KLH conjugated synthetic peptide between 532-561 amino acids from the C-terminal region of human ZDHHC8. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZDHHC8 | |
Probable palmitoyltransferase ZDHHC8, Zinc finger DHHC domain-containing protein 8, DHHC-8, Zinc finger protein 378, ZDHHC8, KIAA1292, ZDHHCL1, ZNF378 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27801, 29801 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction