Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550949

Catalog No. PA550949


Add to cart

Description

Description

ZDHHC9 Polyclonal Antibody detects endogenous levels of ZDHHC9 protein.

The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
Specifications

Specifications

ZDHHC9
Polyclonal
Unconjugated
ZDHHC9
Liquid
ZDHHC9
Synthesized peptide derived from the C-terminal region of human DHHC-9.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y397, P59268
CXorf11, ZDHHC10, ZNF379, ZNF380
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
208884, 302808, 51114
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit