Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zearalenone, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Mycotoxin produced by several species of Fusarium fungi. Binds to the estrogen receptor (ER) (IC50 values are 166 and 240 nM for ERβ and ERα respectively) and stimulates the transcriptional activity of both subtypes at concentrations between 1 - 10 nM. Also stimulates growth of T47D breast cancer cells.
Specifications
Quantity | 50 mg |
Formula Weight | 318.36 |
Percent Purity | >98% |
Chemical Name or Material | Zearalenone |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction