Learn More
ZEB1 Mouse anti-Human, Clone: OTI8A2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802302
Catalog No. 50-166-5936
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
ZEB1
Monoclonal
Unconjugated
ZEB1
Lyophilized
ZEB1
Human recombit protein fragment corresponding to amino acids 1-350 of human ZEB1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8A2
PBS with 8% trehalose and no preservative; pH 7.3
P37275
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6935