Learn More
ZEB1 Rabbit anti-Human, Primate, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP213159AF350
Catalog No. NP21319A30
Add to cart
Description
Description
ZEB1 Polyclonal antibody specifically detects ZEB1 in Human,Primate samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
ZEB1
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ZEB1
A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [Uniprot: P37275]
0.1 mL
Primary
6935
Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot, Immunocytochemistry/Immunofluorescence
AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Primate