Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns, 40K MWCO, 5mL
GSA_VA

Effectively remove salt and other molecules (40K MWCO) from samples and attain excellent protein recovery with these centrifuge-based chromatography columns.

$86.70 - $372.50

Specifications

MWCO 40 kDa
View More Specs

Products 2
Catalog Number Mfr. No. SufficientFor Price Quantity & Availability  
Catalog Number Mfr. No. SufficientFor Price Quantity & Availability  
PI87771
GSA_VA
View Documents Promotion Details
Thermo Scientific™
87771
Pack of 25 for $372.50
Add to cart
 
PI87770
View Documents Promotion Details
Thermo Scientific™
87770
Pack of 5 for $86.70
Add to cart
 
Description

Description

Thermo Scientific™ Zeba Spin Desalting Columns (40K MWCO) contain a size-exclusion chromatography resin that provides excellent protein desalting performance in a centrifuge format for 5μL to 4mL samples.

These specially designed polypropylene centrifuge columns are pre-packed with high-performance Zeba Desalting Resin to facilitate rapid protein desalting with high recovery. The resin effectively traps salts and other small sample-components but does not impede proteins and other macromolecules, which pass directly into the centrifuge collection tube. These particular columns use Zeba Resin having a molecular-weight cutoff (MWCO) rating equal to 40K; this ensures that proteins larger than 40,000 MW will be recovered with high efficiency.

The easy-to-use spin column format dramatically improves results over standard drip-column methodologies. This system eliminates the need to wait for samples to emerge by gravity flow and to monitor fractions for protein recovery. Samples are processed in these desalting columns using a standard centrifuge or microcentrifuge. No chromatography hardware or column preparation steps are required. Multiple samples can be processed in less than ten minutes. Zeba Spin Desalting Columns are available in five different column sizes, and these provide trouble-free desalting and buffer exchange for sample volumes ranging from 5μL to 4mL.

Highlights:

  • Fast centrifuge format – resin and columns are optimized for rapid processing by centrifugation with standard collection tubes and rotors

  • Replaces drip-column methods – no need to collect and screen slowly-emerging protein fractions

  • Exceptional protein yields – recover greater than 95% in typical applications with minimal dilution

  • Five convenient sizes – columns with 75μL to 10mL of Zeba Resin provide desalting matched for sample volumes between 5μL and 4mL

  • 40K MWCO – resin is rated to recover proteins larger than 40,000 daltons and retain (remove) solutes smaller than 2000 MW

Requires:

Microcentrifuge and microcentrifuge collection tubes (for 75μL and 0.5mL columns) or centrifuge and collection tubes (15mL tubes for 2 and 5mL columns; 50mL tubes for 10mL columns)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

40 kDa
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit