Learn More
Thermo Scientific™ Zeba™ Spin Desalting Columns, 7K MWCO, 10 mL Available on GSA/VA Contract for Federal Government customers only.
Effectively remove salt and other molecules (7K MWCO) from samples and attain excellent protein recovery with these centrifuge-based chromatography columns.
$107.00 - $430.00
Products 2
Catalog Number Mfr. No. Sufficient For Quantity Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Sufficient For Quantity Size (Metric) Price Quantity & Availability  
PI89893
View Documents
Thermo Scientific™
89893
5 samples, each 700uL to 4mL 5 columns 10mL
Pack of 5 for $107.00
Add to cart
PI89894 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
89894
25 samples, each 700uL to 4mL 25 columns 10mL
Pack of 25 for $430.00
Add to cart
Description
Description
Thermo Scientific™ Zeba Spin Desalting Columns (7K MWCO) contain a size-exclusion chromatography resin that provides excellent protein desalting performance in a centrifuge format for 2µL to 4mL samples.
These specially designed polypropylene centrifuge columns are pre-packed with high-performance Zeba Desalting Resin to facilitate rapid protein desalting with high recovery. The resin effectively traps salts and other small sample components but does not impede proteins and other macromolecules, which pass directly into the centrifuge collection tube. These particular columns use Zeba Resin having a molecular-weight cutoff (MWCO) rating equal to 7K; this ensures that proteins larger than 7000 MW will be recovered with high efficiency.
The easy-to-use spin column format dramatically improves results over standard drip-column methodologies. This system eliminates the need to wait for samples to emerge by gravity flow and to monitor fractions for protein recovery. Samples are processed in these desalting columns using a standard centrifuge or microcentrifuge. No chromatography hardware or column preparation steps are required. Multiple samples can be processed in less than 10 minutes. Zeba Spin Desalting Columns are available in five different column sizes, and these provide trouble-free desalting and buffer exchange for sample volumes ranging from 2µL to 4mL.
Highlights:
  • Fast centrifuge format – resin and columns are optimized for rapid processing by centrifugation with standard collection tubes and rotors
  • Replaces drip-column methods – no need to collect and screen slowly emerging protein fractions
  • Exceptional protein yields – recover greater than 95% in typical applications with minimal dilution
  • Five convenient sizes – columns with 75µL to 10mL of Zeba Resin provide desalting matched for sample volumes between 2µL and 4mL
  • 7K MWCO – resin is rated to recover proteins larger than 7000 daltons and retain (remove) solutes smaller than 1000 MW
Requires:
Microcentrifuge and microcentrifuge collection tubes (for 75µL and 0.5mL columns) or centrifuge and collection tubes (15mL tubes for 2 and 5mL columns; 50mL tubes for 10mL columns)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.