Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zein, purified, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
Plastic drum
9010-66-6
MFCD00062414
9010-66-6
MFCD00062414
Residue after Ignition 2% max. (800°C)
Heavy Metals 20ppm max
Quantity 100g
Packaging Plastic bottle
Merck Index 14, 10119
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aq. solutions ph >12,aq. ethanol,,soluble in ethanol,dmf
Chemical Name or Material Zein, Purified

EINECSNumber : 232-722-9