Learn More
WASP ZEN Cartridge Core High Temp (HT) Products for Science Education
ZEN Cartridge Core High Temp is an extractable steel cartridge without Teflon for print at high temperature using the WASP ZEN Extruder.
$40.00
Specifications
Product Type 3D Printer Accessory
Material Steel
For Use With (Equipment) WASP ZEN Extruder
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
S4131961 Products for Science Education
View Documents
WASP
WU7018
0.4 mm
Each for $40.00
Add to cart
S4131962 Products for Science Education
View Documents
WASP
WU7019
0.7 mm
Each for $40.00
Add to cart
S4131963 Products for Science Education
View Documents
WASP
WU7020
1.2 mm
Each for $40.00
Add to cart
Description
Description
  • Extractable steel cartridge without Teflon
  • For printing at high temperature
  • Suggested for ABS, PA, PC, HIPS, PA+C, ASA, PMMA, and PP
Specifications
Specifications
3D Printer Accessory
Steel
WASP ZEN Extruder