Learn More
WASP ZEN Cartridge Core Low Temp (LT) Products for Science Education
The ZEN Cartridge Core Low Temp is an extractable steel cartridge with Teflon for printing at low temperature using the WASP ZEN Extruder.
$40.00
Specifications
Product Type 3D Printer Accessory
Material Steel
For Use With (Equipment) WASP ZEN Extruder
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Nozzle Diameter (Metric) Price Quantity & Availability  
S4131958 Products for Science Education
View Documents
WASP
WU7012
0.4 mm
Each for $40.00
Add to cart
S4131959 Products for Science Education
View Documents
WASP
WU7013
0.7 mm
Each for $40.00
Add to cart
S4131960 Products for Science Education
View Documents
WASP
WU7014
1.2 mm
Each for $40.00
Add to cart
Description
Description
  • Extractable steel cartridge with Teflon for WASP ZEN Extruder with Teflon
  • For printing at low temperature
  • For use with PLA, PETG, and FLEX
Specifications
Specifications
3D Printer Accessory
Steel
WASP ZEN Extruder