Learn More
Sepax Technologies IncSupplier Diversity Partner (233150-4630) HPLC Column, Sepax, Zenix-C SEC-150, 3 um, 4.6, x, 300, mm DFS Item
Manufacturer:  Sepax Technologies IncSupplier Diversity Partner 2331504630
Analytical and Preparative Size Exclusion from Sepax Technologies, Inc. || Unix™, Unix™-C, Zenix/ Zenix-C, SRT/ SRT-C, SRT-10/ SRT-10C, and Nanofilm are nine SEC product lines developed based on innovative surface coating and particle technologies comprised of uniform, hydrophilic, and neutral nanometer thick films chemically bonded on 1.8 m, 3 m, 5 m, or 10 m high purity and mechanically stabilized silica. Utilizing two unique surface coating technologies - stand-up monolayer bonded on porous silica for Unix™, Zenix, SRT and SRT-10; and lay-down monolayer on porous silica for Unix™-C, Zenix-C, SRT-C, and SRT-10C, Sepax offers surface chemistry choices based on sample type and sample hydrophobicity in order to achieve high resolution performance separation. For full product details, additional dimensions, rush orders, and custom requests, please contact the manufacturer.
Catalog No. NC1467198