Learn More
PIP™ Zenon™ Z12 Foam
Rimless Safety Glasses feature foam padding seal and anti-scratch / anti-fog coating.
$3.98
Specifications
Frame Type Rimless
Frame Color Black
Lens Type Scratch-resistant, Anti-fog
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-148-036
View Documents
PIP™
25001F020
Clear
Each for $3.98
Case of 144 EA for $573.12
Add to cart
19-148-037
View Documents
PIP™
25001F021
Gray
Each for $3.98
Case of 144 EA for $573.12
Add to cart
Description
Description
  • Lightweight, rimless design provides all day user comfort
  • Foam padding design forms a seal to keep out dust and airborne particles
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points
  • Molded nose bridge design to fit the vast majority of users
  • 9.5 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Specifications
Rimless
Black
Scratch-resistant, Anti-fog