Alfa Aesar™ Zeolite ferrierite, ammonium

Manufacturer: Alfa Aesar™ 4588436

Quantity 500g

 View more versions of this product

Catalog No. AA4588436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1318-02-1
    MDL Number MFCD00132601
    Chemical Name or Material Zeolite ferrierite, ammonium

    Specifications

    Physical Form Powder
    Surface Area 400m2/g
    MDL Number MFCD00132601
    CAS 1318-02-1
    Color White
    Quantity 500g
    Avg. Mol. Wt. or Mol. Wt. Range SiO2:Al2O3, 20:1 mole ratio
    Chemical Name or Material Zeolite ferrierite, ammonium

    Description

    • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit