Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Adenovirus Type 41 Culture Fluid

Infectious virus preparation purified by sucrose gradient centrifugation

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration >105U/mL
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
View More Specs

Alert:

Contains live and infectious organisms.


Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-157-643
View Documents
ZeptoMetrix Corporation
0810085CF
Adenovirus type 41 virus infectious culture fluid
Each for $314.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-660
View Documents
ZeptoMetrix Corporation
0810105CF
Chikungunya virus infectious culture fluid
Each for $314.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Adenoviruses are non-enveloped icosahedral viruses that contain a nucleocapsid and doublestranded linear DNA

  • Consist of virus, cells and media taken directly from tissue culture flask
  • Propagated in HEK293A cell line
  • Biosafety Level 2 organism
  • Available in 1.0mL aliquots
  • Each lot assayed for its Tissue Culture Infective Dose (TCID50)
  • Titer:>105 U/mL

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Nucleic acid /molecular testing, limit of detection (LOD) studies, cross-reactivity studies, other viral-based assays

Specifications

Specifications

2
>105U/mL
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1mL
-65°C
ISO, 13485
Solution
Vial
1 Year
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit