ZeptoMetrix Corporation HAV Vaccine Panels Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner 96110272829

 View more versions of this product

Catalog No. 22-156-952

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Host Species 40 year old Caucasian Female
    Product Type HAV vaccine reference panel over 20 Weeks
    Quantity 5 x 1mL
    Storage Requirements -20°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit