Learn More
ZeptoMetrix Corporation HAV Vaccine Panels Available on GSA/VA Contract for Federal Government customers only.
collected from donors that are negative prior to the vaccination and positive after the vaccination
Manufacturer:  ZeptoMetrix CorporationSupplier Diversity Partner 96110272801
 View more versions of this product
Catalog No. 22-156-946
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
25 year old black male
HAV vaccine reference panel over 31 weeks
5 x 1mL
-20°C